Learn More
World Precision Instrument Bone Cutter, 10.5cm, Straight, Strong Tips
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 503252
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
WPI Bone Cutters have short, thick scissor-style blades, which are designed to efficiently cut through bone tissue. Unlike typical scissor blades, these forceps have a more robust, heavier structure to enable effective cutting and shaping of bone during surgical procedures. The ring handles offer a comfortable grip for the surgeon and provide precise control during bone-cutting procedures. These are 10.5 cm (4.13”) long and are available with curved or straight blades, and smooth or serrated tips. The are made of surgical grade stainless steel, which is suitable for chemical sterilization or autoclaving. Clean your surgical instruments promptly after use to ensure a long life.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.