Learn More
Labconco™ Biosafety Cabinet Replacement Trace Odor Filter
SDP
Removes trace nuisance odors from the exhaust air
Supplier:  Labconco™ 3435602
 View more versions of this product
Catalog No. 34-356-02
Add to Cart
Description
Description
  • Trace Odor Filter for Logic+ Type A2 Biosafety Cabinets.
  • Removes trace nuisance odors from the exhaust air of Logic+ Class II, Type A2 Biosafety Cabinets.
  • Contact Labconco for applications involving light-duty chemicals.
Specifications
Specifications
Trace Odor
4 in.
17.4 in.
84.8 cm
5 ft. Enclosure Width
Purifier Logic+ Class II Type A2 Biosafety Cabinets
33.4 in.
10.2 cm
44.2 cm
Purifier Logic+ Class II Type A2 Biosafety Cabinets
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov