Learn More
Labconco™ Biosafety Cabinet Replacement Trace Odor Filter
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Removes trace nuisance odors from the exhaust air
Supplier: Labconco™ 3435601
Description
- Trace Odor Filter for Logic+ Type A2 Biosafety Cabinets.
- Removes trace nuisance odors from the exhaust air of Logic+ Class II, Type A2 Biosafety Cabinets.
- Contact Labconco for applications involving light-duty chemicals.
Specifications
Trace Odor | |
4 in. | |
17.4 in. | |
59.4 cm | |
4 ft. Enclosure Width |
Purifier Logic+ Class II Type A2 Biosafety Cabinets | |
23.4 in. | |
10.2 cm | |
44.2 cm | |
Purifier Logic+ Class II Type A2 Biosafety Cabinets |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov