Learn More
Siemens Healthineers Berichrom Antithrombin III (A) Reagent
Determination of the functional activity of antithrombin III (AT III) in human plasma.
Supplier: Siemens Healthineers 10446673
This item is not returnable.
View return policy
Description
Berichrom Antithrombin III (A) is a chromogenic activity assay for the detection of hereditary or acquired antithrombin deficiency, thrombophilia, and the monitoring of patients undergoing substitution therapy. The heparin cofactor-independent lyophilized reagent uses bovine thrombin substrate and exhibits no interference with anti-FXa anticoagulants, e.g., rivaroxaban.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
150 Tests/Kit |