Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 290" x 165" Flask size: 3l
Supplier: Bellco Glass, Inc. 196583000
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 290" x 165" Flask size: 3l