Learn More
Electron Microscopy Sciences BEEM Capsule Press
SDP
Supplier:  Electron Microscopy Sciences 6992000
Encompass_Preferred
The Capsule Press is designed to facilitate the removal of blocks from the capsule without any damage to the block tip. PLEASE NOTE: FOR BEEM* capsule size #3, an adapter is required.
Catalog No. 50-246-46
Explore available promotions