Learn More
Clontech Labs 3P BD RETRO-X UNIVERSAL PKG SYSTM
Supplier: Clontech Labs 3P 631530
BD Retro-X Universal Packaging System, 8 RXN, Use w/any Retroviral Vector, 100% Cell Transduce, Efficiently Transduce Hard-To-Transfect Cell Lines, Easily Create Stable Cell Lines, Preparation and Storage: -196degreeC