Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barbituric Acid (Laboratory), Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Specifications
Melting Point | 248°C |
Color | Cream |
Linear Formula | NHCONHCOCH2CO |
Formula Weight | 128.09 |
Grade | Laboratory |
Quantity | 100 g |
Packaging | Glass Bottle |
Physical Form | Powder/Solid |
Chemical Name or Material | Barbituric Acid |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction