Learn More
Interscience Lab Inc BagPage + Full-page filter bag, 400mL, 500/pack
Supplier: Interscience Lab Inc 122025
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum qty of 2 per supplier requirements.
BagPage + is a blender bag with a full-screen microperforated filter. Filtration is instantaneous without risks of cross contaminations. A unique welding system on the bag allows pipetting without error. Filter porosity: 280 microns. Compatible with any laboratory blender. Multilayer: reinforced multicoated complex. Rigid and transparent.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.