Learn More
Hach Company Bag, Sterile, WhirlPak, 207 mL, 100/pk
Supplier: Hach Company 2233199
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Complies with Standard Methods and USEPA for drinking water sample collection. Made from poly resins approved by USDA and FDA for food contact. Sterile (certified), disposable, transparent polyethylene bags with trusted "Whirl-Pak" closure. Suitable for liquid or solid sample collection. "Write-on" surface accepts waterproof ink. 24 oz bag perfect for incubating petri dishes in water bath applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.