Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Geobacillus stearothermophilus ATCC™ 7953™
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
Specifications
ATCC® | 7953TM* |
---|---|
Format | Spore Strips |
Product Type | Geobacillus Species |
Target Organism Class | Gram-positive Rods |
Test Type | Qualitative QC |
Catalog Number | Mfr. No. | ATCC® | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ATCC® | Description | Price | Quantity & Availability | ||||
R4851001
|
Thermo Scientific™
R4851001 |
7953TM* | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population |
Pack of 50 for $519.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
R4851002
|
Thermo Scientific™
R4851002 |
7953TM* | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population |
Pack of 50 for $1,504.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
- 10 -6spore population
Specifications
7953TM* | |
Geobacillus Species | |
Qualitative QC |
Spore Strips | |
Gram-positive Rods |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title