Learn More
Biocare Medical, LLC Background Punisher - 25 ml
Supplier: Biocare Medical, LLC BP974H
Background Punisher is a universal blocking reagent used for reducing nonspecific background staining often found with immunohistochemistry. Background Punisher is specifically formulated for superior pH stability and is sodium azide and thimerosal free. It can be used with automated or manual staining protocols.