Learn More
Biocare Medical, LLC Background Eraser
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Biocare Medical, LLC BE965H
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Background Eraser is specifically formulated for superior pH stability and designed to reduce nonspecific background staining often found with immunohistochemistry techniques. It is optimized to work with Biocare 4plus detection systems and is only used when secondary antibodies are derived in goat. It can also be used on an automated staining system.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.