Learn More
INNOVEX BIOSCIENCES BACKGROUND BUSTER 50ML
Supplier: INNOVEX BIOSCIENCES NB30650
Peptide Blocker delivers complete eradication of background staining of animal, human tissues & cells stained by IHC, IF, in situ ( frozen, paraffin) & free floating sections; Short incubation step. A must for Mouse-on-Mouse staining