Learn More
TARGETMOL CHEMICALS INC AVN-101 5MG
Supplier: TARGETMOL CHEMICALS INC T26690
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AVN-101 is a very potent 5-HT7 receptor antagonist (Ki = 153 pM). AVN-101 also exhibits a rather high affinity toward histamine H1 (Ki = 0.58 nM) and adrenergic (alpha)2A (alpha)2B and (alpha)2C (Ki = 0.41-3.6 nM) receptors. AVN-101 shows a good oral bioavailability and facilitated brain-blood barrier permeability low toxicity and reasonable efficacy in animal models of CNS diseases. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.