Learn More
3M™ Avagard™ D Instant Hand Antiseptic with Moisturizers (61% w/w ethyl alcohol) 9221
Avagard™ D Instant Hand Antiseptic provides fast and effective bacterial kill without being hard on skin.
Supplier:  3M™ 7000002749
Catalog No. 19-072-387
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions
This item is not returnable. View return policy
Description
Description
  • Broad-spectrum activity kills harmful bacteria, including antibiotic-resistant strains (in vitro)
  • Proven efficacy - meets CDC, AORN and WHO Guidelines
  • Helps prevent skin breakdown.
  • Dries quickly without leaving hands sticky
  • Compatible with CHG
  • Recommended for repeat use
  • Reference: 1APIC Guideline for Handwashing and Hand Antisepsis in Health Care Settings, 1995. Larson, E.L. 2CDC Guideline for Handwashing and Hospital Environmental Control, 1985. Julia S. Garner; Martin S. Favero, Hospital Infections Program Center for Infectious Diseases, Centers for Disease Control and Prevention
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hand Sanitizer
61% w/w ethyl alcohol
3 oz.
89 mL
Product Suggestions
Product Suggestions