Learn More
TARGETMOL CHEMICALS INC AV-101 5MG
Supplier:  TARGETMOL CHEMICALS INC T26686
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. AV-101 (4-Cl-KYN) a prodrug antagonist at the glycine site of the NMDA receptor has antidepressant activity and reduces levodopa-induced dyskinesia in MPTP monkeys. purity: 98%
Catalog No. 50-257-7065