Learn More
Perkin Elmer US LLC Auto-Ignite FID Add-On Kit - 240 V, Manual Pneumatics
Supplier: Perkin Elmer US LLC N6120166
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Not compatible with the FID detector on the Clarus 690 and 590 GCs. Refer to the Clarus 590/690 consumables reference guide for more details.