Learn More
Cole-Parmer 25 Pound Solid PP Cable/Zip Ties, 8" L, Black; 1000/Pk
Supplier: Cole-Parmer 0683095
Polypropylene (PP) cable ties are ideal for tubing bundles. PP resists a number of chemicals and is UV-resistant.