Learn More
Mystaire™ Aura™ Main Filter
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For Aura™ Series Ductless Fume Hoods. Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Supplier: Mystaire™ AU2816
Description
- Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Primary | |
Filter | |
Multi-purpose |
Aura™ 42, Aura™ 250 and Aura™ 550 Ductless Fume Hood | |
Gas Phase Carbon | |
Main Filter |