Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Audit™ MicroControls™ MicroFD™ Cardiac Markers Control
Tri-level serum-based control
Supplier: Audit™ MicroControls™ K026M6
This item is not returnable.
View return policy
Description
- Simulate human patient samples for monitoring precision of laboratory testing procedures for CK-MB, Myoglobin and Troponin-I
- Freeze-dried
- Size: 6 x 2mL
Specifications
2°C to 8°C | |
Control FD Cardiac Marker | |
3 |
Creatinine Kinase-MB (CK-MB), Myoglobin, Troponin I | |
Simulate Human patient samples for monitoring precision of laboratory testing procedures for CK-MB, Myoglobin and Troponin-I | |
6 x 2 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction