Learn More
AGI NORTH AMERICA INC Syrris Orb benchtop jacketed reactor combines excellent value with unique user-friendly features, supports vessels from 100mL to10L, equipped with 40Ncm stirrer motor
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: AGI NORTH AMERICA INC 10001ARBF
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Due to its modular and compact design, wide vessel and temperature range, Orb can be used in many applications.Included:-Rugged base frame adaptable to multiple vessel sizes -Easy motor lift for convenient disengagement of the stirrer motor-5L jacketed vessel (1L and 2L available separately)-Torque-limited clamp for rapid vessel change-40 Ncm stirrer motor -Oil drain unit allows for mess-free draining of vessel jacket oil-Operating temp range: -40 to 200 deg C (high range of -90 to 250 deg C available with upgrade kit)-Pressure range: Vacuum: ≤50 mbara. Max pressure: 250 mbar -Temperature probe (LEMO)-Oil pipe for reaction vessel to oil drain unit and connectors -Oil pipe tidy for leak-free storage of oil pipes -PTFE 4-blade propeller stirrer with stirrer seal & guide Order informationNot included: appropriate circulator and pair of oil pipes from the reaction vessel to circulator. Oil pipes require M16 x 1.0 female connection and adapter circulator side
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.