Learn More
Perkin Elmer US LLC ASXpress Plus Aqueous Consumables Kit
Supplier: Perkin Elmer US LLC N0810949
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The contents of this kit are described as follows: 1.0 mm ID Carbon Fiber Supported Sample Probe Aqueous 6-port valve Valve Pump Tubing Kit Nut and Ferrule Placement Fitting (10) CTFE ferrules, 1/16 Internal Standard Tee Kit Passive Bubbler Tee PEEK Tubing, Tee, Fittings and Ferrules