Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ASGPR1 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
$1219.00
Specifications
Assay Range | 39.0625 pg/mL |
---|---|
Assay Sensitivity | 39.06-2500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010143
|
Novus Biologicals™
NBP27955215PLATES |
15 Plates |
Each of 1 for $1,219.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ASGPR1.Specifications
39.0625 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ASGPR1. | |
Sandwich ELISA | |
432 | |
Rabbit | |
Storage is content dependent. |
39.06-2500 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
ASGR1 | |
RUO | |
ASGPR1 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title