Learn More
AdipoGen Ascomycin high purity
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: AdipoGen AGCN20420M025
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Chemical. CAS 104987-12-4. Formula C43H69NO12. MW 792. Isolated from Streptomyces sp. Macrolide antibiotic. Ethyl analog of FK506. Strong immunosuppressant. Binds to the FK-506-binding protein FKBP12. This complex inhibits calcineurin protein phosphatase 2B PP2B. Suppresses the production of T helper type 1 Th1 interferon and IL-2 and Th2 IL-4 and IL-10 cytokines in T lymphocytes and preferentially inhibits the activation of mast cells. Anti-inflammatory. Used for topical treatment of inflammatory skin diseases. Potently inhibits anti-IgE-induced histamine and cytokine release and reduces IgE-dependent p38 MAPK activation in human basophils. Inhibits basophil degranulation at the initial phase of allergic skin reactions. Antifungal. Antimalaria compound. Anticonvulsant antiepileptic.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.