Learn More
Apexbio Technology LLC ARCA, 50ul (100 mM). Cas: MFCD: NA. Anti Reverse Cap Analog (ARCA) used to forming Cap 0 structure,3´-O-Me-m7G(5')ppp(5')G
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Apexbio Technology LLC B8175.05
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A critical step in mRNA processing is the addition of a 5 cap structure, a 5'-5' triphosphate linkage between the 5' initiating terminal of the RNA and a guanosine nucleotide. The cap is then methylated enzymatically at the N-7 position of the guanosine to form a mature mCAP. Other sizes are available. Please inqury us for quote.