Learn More
TARGETMOL CHEMICALS INC APLINDORE FUMARATE 5MG
Supplier: TARGETMOL CHEMICALS INC T30098
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Aplindore Fumarate (DAB-452) is a small molecule dopamine D2 receptor partial agonist.Aplindore Fumarate exhibits high affinity for dopamine D2 and D3 receptors and low affinity for dopamine D4 5-hydroxytryptophan (5-HT2) and (alpha)1-adrenergic receptors.Aplindore Fumarate is used in the study of Parkinson's disease and schizophrenia. Fumarate can be used to study Parkinson's and schizophrenia. purity: 100%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.