Learn More
Vector Laboratories Horse Anti-Rabbit, DyLight 488 DI-1094-1.5
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Vector Laboratories DI10941.5
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Horse Anti-Rabbit IgG Antibody (H+L), DyLight 488, 1.5 mg, is an affinity-purified, ultrapure, high-affinity antibody. It is thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 488 to provide the brightest label for fluorescence microscopy. Supplied in solution. Excitation: 493 nm; Emission: 518 nm; Color: Green.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.