Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG1 κ Mouse, R-PE, Clone: MOPC-21, Isotype Control, BD
For low background screening on a variety of mouse and human tissues. anti-IgG1 κ Mouse, R-PE, Clone: MOPC-21, Isotype Control, BD immunoglobulin is a mouse myeloma protein. This antibody is routinely tested by flow cytometric analysis.
Supplier: BD Biosciences 555749
Description
- Host Species: Mouse
- Clone: MOPC-21
- Isotype: IgG1, κ
- Applications: Flow Cytometry, Isotype Control
- Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Flow Cytometry | |
MOPC-21 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
IgG1 κ |
Monoclonal | |
R-PE | |
Mouse | |
100 Tests | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction