Learn More
Glas Col LLC Evaporator
SDP
Supplier:  Glas Col LLC 099A EV9612S
Encompass_Preferred
Evaporator for 96 well, heated block to maximum 100 degrees C, 120V
Catalog No. NC1248495
Explore available promotions