Learn More
United Scientific™ Analytical Brass Weight Box Set of 9
High quality and economically priced weight set.
Supplier: United Scientific™ AWB100
This item is not returnable.
View return policy
Description
- Analytical Weight Box contains nine brass weights (one each of 1 g, 5 g, 10 g, 50 g and 100 g sizes, and two each of 2 g and 20 g sizes), in polished wooden box
- Includes a 1 to 500 mg fractional weight set, 5 mg rider and forceps
Specifications
Weight Set | |
Brass | |
1, 2, 5, 10, 20, 50, 100 g |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Prop 65