Learn More
EZKEM Amperometric Flow Cell Assembly Complete
Supplier: EZKEM 330001
The complete Amperometric Flow Cell kit includes the Amperometric Flowcell, 0.005” Gasket (326791), Reference Electrode (329513 ) and an allen wrench. If upgrading to this flow cell on a FS3000 or FS3100, a new Amperometric Cable (E100047) will be needed for the detector.