Learn More
Glass Expansion Aluminum Skimmer
Supplier: Glass Expansion TF1002AAL
Aluminum Skimmer Cone (1184050). Compatible with Thermo/Finnigan Element 1, 2 and Neptune ICP-MS. Ideal for lowest Ni background and laser ablation applications. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a boron-free platinum skimmer cone, see catalog number 501907714.For high sensitivity applications and low matrix containing samples, we recommend the Nickel X-Skimmer with Copper Core, see catalog number 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.