Learn More
Perkin Elmer US LLC Aluminum Sample Pans with Quartz Covers for Photo-Calometric Analysis
Supplier: Perkin Elmer US LLC B0196858
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Specialty Pans and Covers are ideal for applications that require high temperatures or large sample volumes. Since these pans and covers have an ambient maximum pressure threshold, they do not require a crimper.