Learn More
Chemglass Life Sciences Condenser, Allihn, 530mm, 24/40 Joint, 400mm Jacket Length
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG120607
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Condenser, Allihn, 530mm, 24/40 Joint, 400mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.