Learn More
Electron Microscopy Sciences EMS Alexander Mastoid Bone Chisel Premium, 14 mm, 7" OAL
Supplier: Electron Microscopy Sciences 7799006
Stainless steel bone chisels are used to score, cut, scrape, clean, and sculpt the bone.
Size: 14 mm