Learn More
AIMS™ Small Animal Thermo Controlled Recovery Board
Keeps animals warm after surgery and will not burn or over heat animal
Supplier: AIMS™ TCRP
Description
- Place in animal cage to provide heat during recovery from anesthesia
- Includes three space gels for continuous heat and two space blankets to retain heat
- Thermo Reflector and Turbo Heat Sink to maximize heat transfer
- Non-skid feet for added stability
- Provides over two hours of Thermo-Regulation
- Fits into standard shoe box
- Size: 13.5 L x 7.5 in. W
Specifications
3 Space Gels/4 Space Blankets | |
Acrylic/KOMATEX | |
13.5 x 7.5 in. |