Learn More
Across International Ai P6A 6L/min SST Peristaltic Pump Touchscreen controller ETL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Across International P6A
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A peristaltic pump, also commonly known as a roller pump, is a type of positive displacement pump used for pumping a variety of fluids. The fluid is contained within a flexible tube fitted inside a circular pump casing (though linear peristaltic pumps have been made). A rotor with a number of "rollers", "shoes", "wipers", or "lobes" attached to the external circumference of the rotor compresses the flexible tube.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.