Learn More
Across International Ai 10°C to 25°C 33L/Min 17L Capacity Recirculating Chiller
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Across International WAC1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Ai WAC-1 digital temperature controlled recirculating water chiller with built-in swivel casters, recommended for our IH series induction heaters (see models below). These chillers have built-in stainless steel tank and water pump for durability and rust-free operation. two sets of water manifolds included for immediate use.
This item is not returnable.
View return policy