Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Agarose LM
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
Axygen™ Low melting agarose allows for the recovery of undamaged nucleic acids below their denaturation temperature.
Supplier: Axygen™ AGRLM501
Description
The low gelling temperature ensures that the agarose will be in a liquid state at a temperature range where In-Gel manipulations can be performed without prior extraction of the DNA from the gel slice. Low Melting (LM) Agarose is derivatized by organic synthesis which generates methoxylate groups from the basic agarose structure. Agarose LM is also optimized for preparative electrophoresis and isolation of DNA fragments.- Highest gelling/melting temperatures and gel strength
- Electrophoresis of DNA fragments ≥1000bp
- Suitable for tissue and cell culture and viral plaque assays
- Ideal for preparative electrophoresis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
LM Agarose Gel | |
Tissue and cell culture and viral plaque assays | |
≥76°C | |
Yes |
Yes | |
Agarose | |
10 x 50 g | |
15° to 25°C |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction