Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Adflo™ PAPR and Versaflo™ M-Series Helmet Kit
Features a heavy duty leather shroud and flame resistant helmet cover. 3M™ Adflo™ PAPR and Versaflo™ M-Series Helmet Kit includes a Speedglas™ Welding Shield M-153SG and Li Ion Battery.
Specifications
Certifications/Compliance | ANSI Z89.1 Type I, Class G |
---|---|
Color | Black |
For Use With (Application) | Welding |
Material | Leather |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
19-070-938
|
3M™
7100200577 |
No ADF |
Case of 1 for $2,764.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Able to lift visors in non-hazardous areas to help facilitate communicationsProtection from welding arc (ANSI Z87) plus spark and splatter
- Head top can be used for most applications including arc gouging, plasma cutting, grinding, MIG, TIG, and GMAW weldingProtection from welding arc (ANSI Z87) plus spark and splatter
- When used in conjunction with NIOSH approved air source, is designed to help protect the user from certain airborne contaminants, and certain head, eye, face, and welding hazardsProtection from welding arc (ANSI Z87) plus spark and splatter
- Compatible with 9100XXi ADF with Natural Color Technology
- 3M Catalog# 38-1101-00SW
Specifications
ANSI Z89.1 Type I, Class G | |
Welding | |
1 Ea. | |
Clear |
Black | |
Leather | |
Standard |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title