Learn More
Promega ADCC Reporter Bioassay, F Variant, Core Kit
Provides a biologically relevant and specific mechanism-of-action-based measure of ADCC without the complex workflow and variability inherent in primary cell-based assays. The bioassay also can be used to quantify effects of antibody glycosylation on Fc effector function.
$1501.00 - $6589.00
Specifications
Content And Storage | Varies Among Components |
---|
Includes: RPMI 1640 Medium
Low IgG Serum
Bio-Glo™ Luciferase Assay Buffer
Bio-Glo™ Luciferase Assay Substrate
ADCC Bioassay Effector Cells, F Variant