Learn More
Cole-Parmer Cole-Parmer Adapter, single pin DIN meter to BNC electrode
Supplier: Cole-Parmer 0598321
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors.Order an adapter to ensure your meter and electrode are compatible.