Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Acrylic Brain Matrices
SDP

Stoelting™ Acrylic Brain Matrices are precisely machined and highly polished to ensure reproducible sectioning.

Supplier:  Stoelting™ 51380

 View more versions of this product

Catalog No. 10-000-763


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Rat and mouse brain matrices are available
  • Coronal and sagittal sections at 1 mm intervals
  • All coronal brain matrices have an additional mid-line sagittal cut to aid in the separation of the left and right hemispheres.
  • Mouse brain matrices can also be used on neonatal rat brains. Divisions are 1 mm apart.
Specifications

Specifications

Neuroscience
40 to 75 g
Mouse, Rat
Coronal
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit