Learn More
Abnova™ BCA Protein Quantitation Kit (Colorimetric)
Use for determining the concentration of protein in solution.
Supplier: Abnova™ KA3718
Specifications
0.5 to 30μg | |
BCA Protein Quantitation Kit is used for determining the concentration of protein in solution. | |
Functional Study |
Store the kit at 4°C. | |
Colorimetric | |
Cell Homogenate, Plasma, Serum, Tissue Homogenate |