Learn More
Anton Paar Digital Performance Refractometer – Abbemat 500
Supplier: Anton Paar 97788
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Abbemat 500 refractometers allow non-destructive refractive index and concentration measurements within seconds. Independent of sample properties you can measure liquids, pastes, polymers, solids, turbid, colored, or opaque samples. - Highly precise quality control measurements of refractive index and concentration at a precision of +/-0.00002 nD- In-built application knowhow with over 200 methods for your sample- Fully 21CFR part 11 compliance- Avoids human errors by showing a clear pass/fail result- Connects to other Anton Paar instruments for multiparameter measurementsMeasuring range Refractive index scale (RI) - Range nD 1.26 to 1.72 - Resolution nD ± 0.000001 - Accuracy nD ± 0.00002 Measuring range Brix scale- Range 0 to 100 % - Resolution 0.001 % - Accuracy 0.015 % Temperature- Range 4 to 85 °C- Accuracy ± 0.03 °C - Stability ± 0.002 °CAccessories: NC1818555, NC1818545, NC1818543
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.