Learn More
NOUBIO NouSerum: Full FBS Replacement (300 proteins)
Supplier: NOUBIO NS50GM
NouSerum, a full slaughter free serum replacement that can lower production costs by 50-95%. NouSerum is composed of all ~300 serum proteins, making it a viable substitute for animal serum in cell-based biomanufacturing. Furthermore, it is in FDA GRAS category, eliminating regulatory issues associated with using animal-based products in cell culture. NouSerum is a powder based product to keep your logistic and storage costs low. 50g of NouSerum is the volume equivalent of 500mL FBS.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.