Learn More
Gilson Company Inc 8oz Stock Solution for Sand Equivalent Test
Supplier: Gilson Company Inc SEA102
Stock Solution used for performing the Sand Equivalent Test. It is a concentrated calcium chloride solution which is diluted with distilled water to create the working solution for conducting Sand Equivalent Tests. To create the working solution, mix 85ml of solution for every gallon of distilled water. The concentrated stock solution has a shelf life of only three months once the container is opened.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.