Learn More
Moldex™ 8000 Series Half-Mask Respirators Cartridges
SDP
For use with 8000 series half mask respirators
Supplier:  Moldex™ 8400
 View more versions of this product
Catalog No. 18-106
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Description
Description
  • Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
  • The P100 cartridge can be used for both oil and non-oil based particulates
  • 100% PVC-Free™
Specifications
Specifications
Series 8000 Half-Mask
Product Suggestions
Product Suggestions