Learn More
Moldex™ 8000 Series Half-Mask P100 Pre-Assembled Respirators
SDP
Pre-assembled 8940 series respirators provide an efficiency level of 99.97% or greater against both oil and non-oil based particulate aerosols. Kit includes 8000 series facepiece, P100 filter disks and filter disk holders.
Supplier:  Moldex™ 8942
 View more versions of this product
Catalog No. 19-003-345B
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Explore available promotions
This item is not returnable. View return policy
Description
Description
  • 8000 Series half-mask respirators pre-assembled with P100 particulate filters
  • Soft, durable facepiece and lightweight design with extra-wide sealing
  • Exhalation valve positioned for better communication
  • Cartridges have a snap-in mount for easy replacement and no misthreading
  • Living hinge allows head harness to be folded comfortably around neck when not in use
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Maintenance made simple with very few replacement parts
  • Fiberglass-free filters, no plastic casing to throw away
Specifications
Specifications
ANSI ISEA 110-2003, Section 7.11.1
Gray
Thermoplastic Elastomer
Large
8942
NIOSH 42 CFR Part 84, TC-23C-1223
P100
Pre-Assembled Half-Mask Respirator
8000 Facepiece. 8940 P100 Particulate Filter Disks and 8900 Filter Disk Holders
Product Suggestions
Product Suggestions